Learn More
TARGETMOL CHEMICALS INC DEXAMETHASONE SODIUM PHO 100MG
SDP
Catalog No. 502576511
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 200 mg 500 mg 1000 mg and bulk. Please contact Fisher for quotes. Dexamethasone Sodium Phosphate (Dexadreson) an interleukin receptor modulator has anti-inflammatory and immunosuppressant effects. It belongs to a potent glucocorticoid class of steroid synthetic drugs. purity: 99%
Catalog No. 50-257-6511 Supplier TARGETMOL CHEMICALS INC Supplier No. T6465100MG
Also available in 10 mg 25 mg 50 mg 200 mg 500 mg 1000 mg and bulk. Please contact Fisher for quotes. Dexamethasone Sodium Phosphate (Dexadreson) an interleukin receptor modulator has anti-inflammatory and immunosuppressant effects. It belongs to a potent glucocorticoid class of steroid synthetic drugs. purity: 99%