Learn More
Selleck Chemical LLC Dexamethasone Sodium Phosphate S4028-1g
SDP
Catalog No. 503109918
Encompass_Preferred
Dexamethasone (Dexamethasone 21-phosphate disodium salt) is a potent synthetic member of the glucocorticoid class of steroid drugs and an interleukin receptor modulator that has anti-inflammatory and immunosuppressant effects
Catalog No. 50-310-9918 Supplier Selleck Chemical LLC Supplier No. S40281G
Add to Cart
Add to Cart
Dexamethasone (Dexamethasone 21-phosphate disodium salt) is a potent synthetic member of the glucocorticoid class of steroid drugs and an interleukin receptor modulator that has anti-inflammatory and immunosuppressant effects