Learn More
Medchemexpress LLC Dexamethasone (Standard) | 50-02-2 | MFCD00064136 | 99.0% | 392.46 | 50 MG
SDP
Catalog No. 5000361539
Encompass_Preferred
Dexamethasone (Standard) is an analytical standard used in research and analytical applications. It acts as a glucocorticoid receptor agonist, significantly reducing the expression of CD11b, CD18, and CD62L on neutrophils, and CD11b and CD18 on monocytes. It is highly effective in managing COVID-19 infection and inhibits the production of exosomes containing inflammatory microRNA-155 in lipopolysaccharide-induced macrophage inflammatory responses.
  • Analytical standard for research and analytical applications
  • Glucocorticoid receptor agonist
  • Reduces expression of CD11b, CD18, and CD62L on neutrophils, and CD11b and CD18 on monocytes
  • Effective in managing COVID-19 infection
  • Inhibits production of exosomes containing inflammatory microRNA-155
  • Used in qualitative, quantitative, and methodological research experiments (HPLC, GC, MS)
Catalog No. 50-003-61539 Supplier Medchemexpress LLC Supplier No. HY14648R50MG
Dexamethasone (Standard) is an analytical standard used in research and analytical applications. It acts as a glucocorticoid receptor agonist, significantly reducing the expression of CD11b, CD18, and CD62L on neutrophils, and CD11b and CD18 on monocytes. It is highly effective in managing COVID-19 infection and inhibits the production of exosomes containing inflammatory microRNA-155 in lipopolysaccharide-induced macrophage inflammatory responses.
  • Analytical standard for research and analytical applications
  • Glucocorticoid receptor agonist
  • Reduces expression of CD11b, CD18, and CD62L on neutrophils, and CD11b and CD18 on monocytes
  • Effective in managing COVID-19 infection
  • Inhibits production of exosomes containing inflammatory microRNA-155
  • Used in qualitative, quantitative, and methodological research experiments (HPLC, GC, MS)