Learn More
Medchemexpress LLC Dexamethasone (Water Soluble) | 50-02-2 | 98.9% | 50 MG
SDP
Catalog No. 5000363650
Encompass_Preferred
Dexamethasone (Hexadecadrol) Water Soluble is a water-soluble form of Dexamethasone, acting as a glucocorticoid receptor agonist and apoptosis inducer. It serves as a common disease inducer in experimental animals, suitable for models of muscle atrophy, hypertension, and depression. It also inhibits inflammatory miRNA-155 exosomes and reduces inflammatory factors, with potential in COVID-19 research.
  • Glucocorticoid receptor agonist and apoptosis inducer
  • Used for muscle atrophy, hypertension, and depression models
  • Inhibits inflammatory miRNA-155 exosomes; reduces inflammatory factors
  • Potential for COVID-19 research
  • Solubility in H2O: ≥ 100 mg/mL
  • Modulates key transcription factors
  • Potently inhibits GM-CSF release
Catalog No. 50-003-63650 Supplier Medchemexpress LLC Supplier No. HY14648C50MG
Dexamethasone (Hexadecadrol) Water Soluble is a water-soluble form of Dexamethasone, acting as a glucocorticoid receptor agonist and apoptosis inducer. It serves as a common disease inducer in experimental animals, suitable for models of muscle atrophy, hypertension, and depression. It also inhibits inflammatory miRNA-155 exosomes and reduces inflammatory factors, with potential in COVID-19 research.
  • Glucocorticoid receptor agonist and apoptosis inducer
  • Used for muscle atrophy, hypertension, and depression models
  • Inhibits inflammatory miRNA-155 exosomes; reduces inflammatory factors
  • Potential for COVID-19 research
  • Solubility in H2O: ≥ 100 mg/mL
  • Modulates key transcription factors
  • Potently inhibits GM-CSF release