Learn More
Cayman Chemical Dexchlorpheniramine (maleate), Purity: ≥98%, CAS: 2438-32-6, 1 g
SDP
Catalog No. 502933796
Encompass_Preferred
A histamine H1 receptor antagonist (pA2 = 9.36 in guinea pig ileal tissue); inhibits specific allergen-challenged peripheral blood mononuclear cell proliferation by 92% in vitro (4.8 µM); reduces noradrenaline uptake in rat vas deferens ex vivo (10 µM); increases pain threshold in mice exposed to thermal and chemical stimulation (30 mg/kg, i.p.)
Catalog No. 50-293-3796 Supplier Cayman Chemical Supplier No. 240271 G
A histamine H1 receptor antagonist (pA2 = 9.36 in guinea pig ileal tissue); inhibits specific allergen-challenged peripheral blood mononuclear cell proliferation by 92% in vitro (4.8 µM); reduces noradrenaline uptake in rat vas deferens ex vivo (10 µM); increases pain threshold in mice exposed to thermal and chemical stimulation (30 mg/kg, i.p.)