Learn More
Med Vet International 0.5mg/mL, 10mL Zoetis Dexdomitor, Veterinary Prescription
Catalog No. 502827816
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Dexdomitor (dexmedetomidine hydrochloride 0.5 mg/mL) is a synthetic alpha2-adrenoreceptor agonist with sedative and analgesic properties. It is indicated for use as a sedative and analgesic in dogs and cats to facilitate clinical examinations, clinical procedures, minor surgical procedures, and minor dental procedures. Also indicated for use as a preanesthetic to general anesthesia in dogs. Administer IM or IV.
Catalog No. 50-282-7816 Supplier Med Vet International Supplier No. RXDEXDOMITOR
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Dexdomitor (dexmedetomidine hydrochloride 0.5 mg/mL) is a synthetic alpha2-adrenoreceptor agonist with sedative and analgesic properties. It is indicated for use as a sedative and analgesic in dogs and cats to facilitate clinical examinations, clinical procedures, minor surgical procedures, and minor dental procedures. Also indicated for use as a preanesthetic to general anesthesia in dogs. Administer IM or IV.