Learn More
Medchemexpress LLC HY-105545 10mg Medchemexpress, Dexetimide CAS:21888-98-2 Purity:>98%
SDP
Catalog No. 501932265
Encompass_Preferred
Medchemexpress, HY-105545 10mg Dexetimide CAS:21888-98-2 Dexetimide ((+)-Benzetimide) is a high-affinity muscarinic receptor antagonist and a potent and persistent anticholinergic agent used to treat neuroleptic-induced parkinsonism [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2265 Supplier Medchemexpress LLC Supplier No. HY10554510MG
Medchemexpress, HY-105545 10mg Dexetimide CAS:21888-98-2 Dexetimide ((+)-Benzetimide) is a high-affinity muscarinic receptor antagonist and a potent and persistent anticholinergic agent used to treat neuroleptic-induced parkinsonism [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.