Learn More
MSC North Dexi-Task Disposable Gloves, Nitrile, 5.0 mil, Medium, 100/Pk.
Catalog No. 5000508201
Encompass
These blue, powder-free nitrile gloves are designed for general purpose applications. The ambidextrous, disposable gloves provide a consistent fit and protection in various workplace environments.
  • Material: Nitrile
  • Thickness: 5.0 mil
  • Size: Medium
  • Length: 9.0 in.
  • Finish: Smooth
  • Cuff style: Rolled
  • AQL: 1.5
  • FDA approved for food contact
Catalog No. 50-005-08201 Supplier MSC Supplier No. 54641162
These blue, powder-free nitrile gloves are designed for general purpose applications. The ambidextrous, disposable gloves provide a consistent fit and protection in various workplace environments.
  • Material: Nitrile
  • Thickness: 5.0 mil
  • Size: Medium
  • Length: 9.0 in.
  • Finish: Smooth
  • Cuff style: Rolled
  • AQL: 1.5
  • FDA approved for food contact