Learn More
Selleck Chemical LLC Dexketoprofen trometamol E4807-1G
SDP
Catalog No. 5000043319
Encompass_Preferred
Dexketoprofen trometamol (Dexketoprofen tromethamine salt) is an orally active non-selective inhibitor of COX Dexketoprofen trometamol exhibits analgesic efficacy and can be used in treating acute pain
Catalog No. 50-000-43319 Supplier Selleck Chemical LLC Supplier No. E48071G
Dexketoprofen trometamol (Dexketoprofen tromethamine salt) is an orally active non-selective inhibitor of COX Dexketoprofen trometamol exhibits analgesic efficacy and can be used in treating acute pain