Learn More
Apexbio Technology LLC Dexlansoprazole 138530-94-6 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061971
Encompass_Preferred
Dexlansoprazole (CAS 138530-94-6) is the dextrorotatory isomer of lansoprazole classified pharmacologically as a proton pump inhibitor (PPI) It exerts its pharmacological effects by selectively inhibiting the gastric H /K -ATPase enzyme system thereby reducing gastric acid secretion Its dual delayed-release formulation facilitates prolonged acid suppression which has been investigated extensively for therapeutic utility in acid-related gastrointestinal disorders including gastroesophageal reflux disease (GERD) erosive esophagitis and associated conditions
Catalog No. 50-000-61971 Supplier Apexbio Technology LLC Supplier No. B17255.1
Dexlansoprazole (CAS 138530-94-6) is the dextrorotatory isomer of lansoprazole classified pharmacologically as a proton pump inhibitor (PPI) It exerts its pharmacological effects by selectively inhibiting the gastric H /K -ATPase enzyme system thereby reducing gastric acid secretion Its dual delayed-release formulation facilitates prolonged acid suppression which has been investigated extensively for therapeutic utility in acid-related gastrointestinal disorders including gastroesophageal reflux disease (GERD) erosive esophagitis and associated conditions