Learn More
Apexbio Technology LLC Dexlansoprazole,50mg CAS# 138530-94-6
SDP
Catalog No. 502240607
Encompass_Preferred
Dexlansoprazole, the dextrorotatory enantiomer of lansoprazole, is a proton pump inhibitor (PPI) formulated to have dual delayed-release properties. Other sizes are also available. Please inqury us for quote.
This item is not returnable. View return policy
Catalog No. 50-224-0607 Supplier Apexbio Technology LLC Supplier No. B172550
Dexlansoprazole, the dextrorotatory enantiomer of lansoprazole, is a proton pump inhibitor (PPI) formulated to have dual delayed-release properties. Other sizes are also available. Please inqury us for quote.