Learn More
Cayman Chemical Dexloxiglumide, Purity: ≥95%, CAS: 119817-90-2, 10 mg
SDP
Catalog No. 502910875
Encompass_Preferred
A CCK1 receptor antagonist; selectively binds to the CCK1 receptor over CCK2 in CHO cells expressing the rat receptors (Kis = 0.0234 and 5.88 µM, respectively); inhibits guinea pig CCK-8S-induced gall bladder contractions at 20 µmol/kg; reduces CCK-8-induced gastric emptying in rats (ID50 = 1.14 mg/kg)
Catalog No. 50-291-0875 Supplier Cayman Chemical Supplier No. 3475910 MG
A CCK1 receptor antagonist; selectively binds to the CCK1 receptor over CCK2 in CHO cells expressing the rat receptors (Kis = 0.0234 and 5.88 µM, respectively); inhibits guinea pig CCK-8S-induced gall bladder contractions at 20 µmol/kg; reduces CCK-8-induced gastric emptying in rats (ID50 = 1.14 mg/kg)