Learn More
Medchemexpress LLC HY-128878 1mg Medchemexpress, Dexloxiglumide CAS:119817-90-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1288781MG
Encompass_Preferred
Medchemexpress, HY-128878 1mg Dexloxiglumide CAS:119817-90-2 Dexloxiglumide is a selective cholecystokinin type A (CCKA) receptor antagonist [1] . Dexloxiglumide, the active enantiomer of Loxiglumide, inhibits smooth muscle cell contractions induced by cholecystokinin-octapeptide (CCK-8) [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2643