Learn More
Med Vet International Dexmedetomidine Hcl Injection, 0.5 mg/mL, 10mL
Catalog No. 502827811
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Dexmedetomidine HCL is indicated for use as a sedative and analgesic in dogs and cats to facilitate clinical examinations, clinical procedures, minor surgical procedures, and minor dental procedures. Dexmedetomidine HCL is also indicated for use as a pre-anesthetic to general anesthesia in dogs and cats.
Catalog No. 50-282-7811 Supplier Med Vet International Supplier No. RXDEXMEDHCL
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Dexmedetomidine HCL is indicated for use as a sedative and analgesic in dogs and cats to facilitate clinical examinations, clinical procedures, minor surgical procedures, and minor dental procedures. Dexmedetomidine HCL is also indicated for use as a pre-anesthetic to general anesthesia in dogs and cats.