Learn More
Cayman Chemical Dexmedetomidine, Purity: ≥98%, CAS: 113775-47-6, 50 mg
SDP
Catalog No. 502916204
Encompass_Preferred
An α2-AR agonist; selectively binds to α2-ARs over α1-ARs (Kis = 0.015 and 5 nM, respectively); prevents increases in CTnI, as well as reduces myocardial apoptosis, pyroptosis, and ferroptosis in a mouse model of CLP-induced sepsis at 50 µg/kg; induces sedation and decreases mechanical nociception in cats at 40 µg/kg
Catalog No. 50-291-6204 Supplier Cayman Chemical Supplier No. 3954450 MG
An α2-AR agonist; selectively binds to α2-ARs over α1-ARs (Kis = 0.015 and 5 nM, respectively); prevents increases in CTnI, as well as reduces myocardial apoptosis, pyroptosis, and ferroptosis in a mouse model of CLP-induced sepsis at 50 µg/kg; induces sedation and decreases mechanical nociception in cats at 40 µg/kg