Learn More
Medchemexpress LLC Dexmedetomidine | 113775-47-6 | 99.9% | 50 MG
SDP
Catalog No. 5000341147
Encompass_Preferred
Dexmedetomidine is a potent, selective, and orally active agonist of α2-adrenoceptor, with a Ki of 1.08 nM. It shows 1620-fold selectivity against α1-adrenoceptor and exhibits anxiolysis, sedation, and modest analgesia effects.
  • Potent α2-adrenoceptor agonist
  • Selective α2-adrenoceptor agonist
  • Orally active
  • Exhibits anxiolysis
  • Provides sedation
  • Offers modest analgesia effects
Catalog No. 50-003-41147 Supplier Medchemexpress LLC Supplier No. HY1271950MG
Dexmedetomidine is a potent, selective, and orally active agonist of α2-adrenoceptor, with a Ki of 1.08 nM. It shows 1620-fold selectivity against α1-adrenoceptor and exhibits anxiolysis, sedation, and modest analgesia effects.
  • Potent α2-adrenoceptor agonist
  • Selective α2-adrenoceptor agonist
  • Orally active
  • Exhibits anxiolysis
  • Provides sedation
  • Offers modest analgesia effects