Learn More
TARGETMOL CHEMICALS INC Dexmedetomidine 200MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T2524200MG
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Dexmedetomidine is a Central alpha-2 Adrenergic Agonist. The mechanism of action of dexmedetomidine is as an Adrenergic alpha2-Agonist. The physiologic effect of dexmedetomidine is by means of General Anesthesia. Purity 99.96%
Catalog No. 50-287-7422