Learn More
AdipoGen Dexmedetomidine hydrochloride (10 mg)
SDP
Supplier:  AdipoGen CDXD0792M010
Encompass_Preferred
Chemical. CAS: 145108-58-3. Formula: C13H16N2 · HCl. MW: 236.74. Dexmedetomidine hydrochloride is the active isomer of medetomidine and is a highly potent and selective alpha2-adrenergic agonist (Ki values are 1.08 and 1750 nM for alpha2- and alpha1-adrenoceptors respectively) with analgesic and sedative properties. Dexmedetomidine hydrochloride is used clinically as an anesthetic agent to reduce anxiety and tension and promote relaxation and sedation without causing respiratory depression. It is active in vivo; displays hypotensive, bradycardic, sedative, anxiolytic, hypothermic and analgesic effects.
Catalog No. 50-205-4399