Learn More
Apexbio Technology LLC Dexmedetomidine HCl 145108-58-3 10mM (in 1mL DMSO)
SDP
Catalog No. 5000092541
Encompass_Preferred
Dexmedetomidine Hydrochloride (CAS 145108-58-3) is a selective agonist of the adrenergic alpha-2 receptor By binding to these presynaptic receptors it inhibits the release of norepinephrine resulting in decreased sympathetic activity Dexmedetomidine HCl is widely utilized in biomedical research to investigate mechanisms of analgesia sedation and autonomic regulation It is also employed in veterinary studies for its ability to induce sedation and analgesia supporting research on pain pathways sedation protocols and alpha-2 adrenergic pharmacology
Catalog No. 50-000-92541 Supplier Apexbio Technology LLC Supplier No. B13575.1
Dexmedetomidine Hydrochloride (CAS 145108-58-3) is a selective agonist of the adrenergic alpha-2 receptor By binding to these presynaptic receptors it inhibits the release of norepinephrine resulting in decreased sympathetic activity Dexmedetomidine HCl is widely utilized in biomedical research to investigate mechanisms of analgesia sedation and autonomic regulation It is also employed in veterinary studies for its ability to induce sedation and analgesia supporting research on pain pathways sedation protocols and alpha-2 adrenergic pharmacology