Learn More
TARGETMOL CHEMICALS INC DEXMEDETOMIDINE HYDROCHL 50MG
SDP
Catalog No. 502579356
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Dexmedetomidine hydrochloride (Precedex) is an imidazole derivative that is an agonist of the adrenergic alpha-2 receptor. It is closely-related to MEDETOMIDINE which is the racemic form of this compound. purity: 99%
Catalog No. 50-257-9356 Supplier TARGETMOL CHEMICALS INC Supplier No. T646650MG
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Dexmedetomidine hydrochloride (Precedex) is an imidazole derivative that is an agonist of the adrenergic alpha-2 receptor. It is closely-related to MEDETOMIDINE which is the racemic form of this compound. purity: 99%