Learn More
Cayman Chemical DexmedetomIdInhydrochLRId 10mg
SDP
Catalog No. 502971161
Encompass_Preferred
The (S)-enantiomer of medetomidine, an imidazole compound that binds to α2-adrenoceptors (Ki = 1.08 nM) with high affinity and selectivity; its activation of α2-adrenoceptors produces analgesic, sedative, and anxiolytic effects
Catalog No. 50-297-1161 Supplier Cayman Chemical Supplier No. 1558110 MG
The (S)-enantiomer of medetomidine, an imidazole compound that binds to α2-adrenoceptors (Ki = 1.08 nM) with high affinity and selectivity; its activation of α2-adrenoceptors produces analgesic, sedative, and anxiolytic effects