Learn More
Cayman Chemical Dexmedetomidine (hydrochloride), Purity: ≥99%, CAS: 145108-58-3, 50 mg
SDP
Catalog No. 502969371
Encompass_Preferred
The (S)-enantiomer of medetomidine, an imidazole compound that binds to α2-adrenoceptors (Ki = 1.08 nM) with high affinity and selectivity; its activation of α2-adrenoceptors produces analgesic, sedative, and anxiolytic effects
Catalog No. 50-296-9371 Supplier Cayman Chemical Supplier No. 1558150 MG
The (S)-enantiomer of medetomidine, an imidazole compound that binds to α2-adrenoceptors (Ki = 1.08 nM) with high affinity and selectivity; its activation of α2-adrenoceptors produces analgesic, sedative, and anxiolytic effects