Learn More
TARGETMOL CHEMICALS INC Dexpramipexole dihydrochloride
SDP
Catalog No. 502878556
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Dexpramipexole dihydrochloride (KNS-760704 dihydrochloride) ((R)-Pramipexole dihydrochloride) is a neuroprotective agent and is a weak non-ergoline dopamine agonist. Purity 99.88%
Catalog No. 50-287-8556 Supplier TARGETMOL CHEMICALS INC Supplier No. T1101125MG
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Dexpramipexole dihydrochloride (KNS-760704 dihydrochloride) ((R)-Pramipexole dihydrochloride) is a neuroprotective agent and is a weak non-ergoline dopamine agonist. Purity 99.88%