Learn More
MSC Honeywell Dexpure Nitrile Exam Gloves, Non-Sterile, Large, 200/Box
Catalog No. 5000508671
Encompass
These lightweight, powder-free, and latex-free nitrile exam gloves are designed for patient care, providing the comfort and sensitivity of natural rubber latex. These ambidextrous, non-sterile gloves offer excellent tactile sensitivity and dexterity for various healthcare settings.
  • Textured fingertips for enhanced grip
  • Beaded cuff for ease of donning
  • Compliant with EN455, EN374, ASTM D6319, and ASTM D6978 standards
  • Manufactured in accordance with ISO 13485
  • Cleared for use with various chemotherapy drugs
  • Outstanding tactile sensitivity with 0.09 mm finger thickness
  • Tensile strength of 27 MPa before aging
  • Breakthrough time of > 240 minutes for most tested chemotherapy drugs
Catalog No. 50-005-08671 Supplier MSC Supplier No. 21025416
These lightweight, powder-free, and latex-free nitrile exam gloves are designed for patient care, providing the comfort and sensitivity of natural rubber latex. These ambidextrous, non-sterile gloves offer excellent tactile sensitivity and dexterity for various healthcare settings.
  • Textured fingertips for enhanced grip
  • Beaded cuff for ease of donning
  • Compliant with EN455, EN374, ASTM D6319, and ASTM D6978 standards
  • Manufactured in accordance with ISO 13485
  • Cleared for use with various chemotherapy drugs
  • Outstanding tactile sensitivity with 0.09 mm finger thickness
  • Tensile strength of 27 MPa before aging
  • Breakthrough time of > 240 minutes for most tested chemotherapy drugs