Learn More
Apexbio Technology LLC Dexrazoxane 24584-09-6 10mg
SDP
Catalog No. 502240469
Encompass_Preferred
Dexrazoxane is a cell-permeable iron chelator that suppresses intracellular superoxide radical generation via iron sequestration thereby exhibiting antioxidant activity Dexrazoxane exerts its biological effects primarily by modulating oxidative stress and ferroptosis pathways In cell-based studies including myocardial H9c2 cell models dexrazoxane inhibits ferroptosis by suppressing the HMGB1 pathway and induces DNA damage and apoptosis in various tumor cell lines Based on these pharmacological properties dexrazoxane holds research potential in the study of cardioprotective mechanisms tumor-related DNA damage responses ferroptosis and oxidative stress processes
This item is not returnable. View return policy
Catalog No. 50-224-0469 Supplier Apexbio Technology LLC Supplier No. B340710
Dexrazoxane is a cell-permeable iron chelator that suppresses intracellular superoxide radical generation via iron sequestration thereby exhibiting antioxidant activity Dexrazoxane exerts its biological effects primarily by modulating oxidative stress and ferroptosis pathways In cell-based studies including myocardial H9c2 cell models dexrazoxane inhibits ferroptosis by suppressing the HMGB1 pathway and induces DNA damage and apoptosis in various tumor cell lines Based on these pharmacological properties dexrazoxane holds research potential in the study of cardioprotective mechanisms tumor-related DNA damage responses ferroptosis and oxidative stress processes