Learn More
Selleck Chemical LLC Dexrazoxane
SDP
Catalog No. 5000003200
Encompass_Preferred
Dexrazoxane (ICRF-187) is an anticancer drug able to block mitosis and to arrest dividing cells It is an iron chelator and provides cardioprotection against anthracycline toxicity
Catalog No. 50-000-03200 Supplier Selleck Chemical LLC Supplier No. S5651100MG
Dexrazoxane (ICRF-187) is an anticancer drug able to block mitosis and to arrest dividing cells It is an iron chelator and provides cardioprotection against anthracycline toxicity