Learn More
Apexbio Technology LLC Dexrazoxane 24584-09-6 100mg
SDP
Supplier:  Apexbio Technology LLC B3407100
Encompass_Preferred
Dexrazoxane is a cell-permeable iron chelator that suppresses intracellular superoxide radical generation via iron sequestration thereby exhibiting antioxidant activity Dexrazoxane exerts its biological effects primarily by modulating oxidative stress and ferroptosis pathways In cell-based studies including myocardial H9c2 cell models dexrazoxane inhibits ferroptosis by suppressing the HMGB1 pathway and induces DNA damage and apoptosis in various tumor cell lines Based on these pharmacological properties dexrazoxane holds research potential in the study of cardioprotective mechanisms tumor-related DNA damage responses ferroptosis and oxidative stress processes
Catalog No. 50-000-94108