Learn More
Cayman Chemical Dexrazoxane, Purity: ≥98%, CAS: 24584-09-6, 25 mg
SDP
Catalog No. 502968999
Encompass_Preferred
An intracellular iron chelator and an inhibitor of topoisomerase IIα that is highly protective in reducing anthracycline-induced cardiotoxicity and extravasation injury; scavenges hydroxyl (IC50 = 2.1 mM), superoxide (IC50 = 0.4 mM), lipid (IC50 = 4.4 mM), DPPH (IC50 = 3.5 μM), and ABTS+ (IC50 = 3.8 mM) free radicals
Catalog No. 50-296-8999 Supplier Cayman Chemical Supplier No. 1463225 MG
An intracellular iron chelator and an inhibitor of topoisomerase IIα that is highly protective in reducing anthracycline-induced cardiotoxicity and extravasation injury; scavenges hydroxyl (IC50 = 2.1 mM), superoxide (IC50 = 0.4 mM), lipid (IC50 = 4.4 mM), DPPH (IC50 = 3.5 μM), and ABTS+ (IC50 = 3.8 mM) free radicals