Learn More
TARGETMOL CHEMICALS INC DEXRAZOXANE 500MG
SDP
Catalog No. 502580965
Encompass_Preferred
Also available in 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Dexrazoxane (ICRF-187) is an anticancer drug used to block mitosis and arrest dividing cells. It is an iron chelator and provides cardioprotection against anthracycline toxicity. purity: 99%
Catalog No. 50-258-0965 Supplier TARGETMOL CHEMICALS INC Supplier No. T22305500MG
Also available in 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Dexrazoxane (ICRF-187) is an anticancer drug used to block mitosis and arrest dividing cells. It is an iron chelator and provides cardioprotection against anthracycline toxicity. purity: 99%