Learn More
Selleck Chemical LLC Dexrazoxane HCl (ICRF-187) 10mg 149003-01-0 ADR-529
SDP
Supplier:  Selleck Chemical LLC S122210MG
Encompass_Preferred
Dexrazoxane HCl (ICRF-187, ADR-529) is an intracellular iron chelator, which decreases the formation of superoxide radicals, used as a cardioprotective agent; also an inhibitor of topoisomerase II *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-624-9