Learn More
Medchemexpress LLC Dexrazoxane hydrochloride | 149003-01-0 | >99.8% | C11H18Cl2N4O4 | 50 MG
SDP
Catalog No. 5000434187
Encompass_Preferred
Dexrazoxane hydrochloride is a heart protectant with antioxidant and anti-inflammatory properties. It can cross the blood-brain barrier, improving motor function disorders and offering neuroprotective effects, making it useful in the study of neurodegenerative diseases. As a derivative of EDTA, it chelates iron, reducing the formation of superoxide radicals and protecting the heart against the cardiotoxic side effects of anthracyclines like doxorubicin.
  • Heart protectant with antioxidant and anti-inflammatory properties
  • Crosses the blood-brain barrier, improving motor function disorders
  • Offers neuroprotective effects, useful in neurodegenerative disease studies
  • Chelates iron, reducing superoxide radical formation
  • Protects the heart against cardiotoxic side effects of anthracyclines
Catalog No. 50-004-34187 Supplier Medchemexpress LLC Supplier No. HY7620150MG
Dexrazoxane hydrochloride is a heart protectant with antioxidant and anti-inflammatory properties. It can cross the blood-brain barrier, improving motor function disorders and offering neuroprotective effects, making it useful in the study of neurodegenerative diseases. As a derivative of EDTA, it chelates iron, reducing the formation of superoxide radicals and protecting the heart against the cardiotoxic side effects of anthracyclines like doxorubicin.
  • Heart protectant with antioxidant and anti-inflammatory properties
  • Crosses the blood-brain barrier, improving motor function disorders
  • Offers neuroprotective effects, useful in neurodegenerative disease studies
  • Chelates iron, reducing superoxide radical formation
  • Protects the heart against cardiotoxic side effects of anthracyclines