Learn More
Medchemexpress LLC Dexrazoxane (hydrochloride) | 149003-01-0 | 99.83% | C11H18Cl2N4O4 | 1 ML
SDP
Catalog No. 5000434638
Encompass_Preferred
Dexrazoxane hydrochloride, also known as ICRF-187 hydrochloride, is a heart protectant that helps preserve ovarian function and fertility. It exhibits antioxidant and anti-inflammatory properties, can cross the blood-brain barrier, and offers neuroprotective effects for neurodegenerative disease studies. Functioning as an iron chelator, it reduces anthracycline-induced cardiotoxicity by decreasing the formation of superoxide radicals.
  • Preserves ovarian function and fertility.
  • Possesses antioxidant and anti-inflammatory properties.
  • Crosses the blood-brain barrier.
  • Improves motor function disorders.
  • Offers neuroprotective effects.
  • Chelates iron to reduce cardiotoxicity from anthracyclines.
  • Induces DNA breaks and apoptosis in human fibrosarcoma cell lines.
  • Protects ovarian cells from DXR-induced DNA damage.
Catalog No. 50-004-34638 Supplier Medchemexpress LLC Supplier No. HY7620110MM/1ML
Dexrazoxane hydrochloride, also known as ICRF-187 hydrochloride, is a heart protectant that helps preserve ovarian function and fertility. It exhibits antioxidant and anti-inflammatory properties, can cross the blood-brain barrier, and offers neuroprotective effects for neurodegenerative disease studies. Functioning as an iron chelator, it reduces anthracycline-induced cardiotoxicity by decreasing the formation of superoxide radicals.
  • Preserves ovarian function and fertility.
  • Possesses antioxidant and anti-inflammatory properties.
  • Crosses the blood-brain barrier.
  • Improves motor function disorders.
  • Offers neuroprotective effects.
  • Chelates iron to reduce cardiotoxicity from anthracyclines.
  • Induces DNA breaks and apoptosis in human fibrosarcoma cell lines.
  • Protects ovarian cells from DXR-induced DNA damage.