Learn More
Cayman Chemical DeXromethorphnhydrobromId 50mg
SDP
Catalog No. 502954477
Encompass_Preferred
An analytical reference standard categorized as an antitussive; has analgesic and anticonvulsant activities in rodents; has been found as an adulterant in 3,4-MDMA; has potential for abuse; intended for research and forensic applications
Catalog No. 50-295-4477 Supplier Cayman Chemical Supplier No. 1395050 MG
An analytical reference standard categorized as an antitussive; has analgesic and anticonvulsant activities in rodents; has been found as an adulterant in 3,4-MDMA; has potential for abuse; intended for research and forensic applications