Learn More
Grainger Superior Glove Dexterity Knit Gloves, Polyurethane Palm, Cut Level A4, Size 6, 1 Pair
Catalog No. 502780034
Encompass_Preferred
These gloves feature an engineered ParaActiv™ yarn liner, providing 360° cut protection with a bare hand feel. The smooth, non-marring polyurethane palm dip offers enhanced dry grip without leaving residue, making them ideal for delicate handling tasks.
  • ANSI A4 cut resistance
  • ANSI 4 abrasion resistance and ANSI 2 puncture resistance
  • Engineered for a close, snug ergonomic fit
  • Non-marring coating suitable for glass handling
  • Fine knit construction for tactile sensitivity
  • Suitable for use in aerospace, automotive, glass manufacturing, and metal fabrication
  • Washable in hot water and safe for tumble drying on high heat
Catalog No. 50-278-0034 Supplier Grainger Supplier No. 19NE05
These gloves feature an engineered ParaActiv™ yarn liner, providing 360° cut protection with a bare hand feel. The smooth, non-marring polyurethane palm dip offers enhanced dry grip without leaving residue, making them ideal for delicate handling tasks.
  • ANSI A4 cut resistance
  • ANSI 4 abrasion resistance and ANSI 2 puncture resistance
  • Engineered for a close, snug ergonomic fit
  • Non-marring coating suitable for glass handling
  • Fine knit construction for tactile sensitivity
  • Suitable for use in aerospace, automotive, glass manufacturing, and metal fabrication
  • Washable in hot water and safe for tumble drying on high heat