Learn More
Cayman Chemical Dextra sulfatesodIum salt 25g
SDP
Catalog No. 502986852
Encompass_Preferred
A sulfated polysaccharide; induces colonic inflammation in a mouse model of IBD; acute, chronic, and relapsing models of IBD can be induced by varying the concentration and administration frequency of dextran sulfate; has been used in molecular biology applications including precipitation of lipoproteins, acceleration of DNA probe hybridization to nucleic acids, and ribosomal isolations
Catalog No. 50-298-6852 Supplier Cayman Chemical Supplier No. 2325025 G
A sulfated polysaccharide; induces colonic inflammation in a mouse model of IBD; acute, chronic, and relapsing models of IBD can be induced by varying the concentration and administration frequency of dextran sulfate; has been used in molecular biology applications including precipitation of lipoproteins, acceleration of DNA probe hybridization to nucleic acids, and ribosomal isolations