Learn More
Immudex MHC Dextramer, 500 tests, 10 μl per test, HLA-A*02:01, SLLMWITQV peptide, NY-ESO-1 specificity, BV421, FITC, PE, APC available
SDP
Supplier:  Immudex WB03247 AP 500
Encompass
MHC Dextramer, 500 tests, 10 μl per test, HLA-A*02:01, SLLMWITQV peptide, NY-ESO-1 specificity, BV421, FITC, PE, APC available
Catalog No. NC3667574
Only null left
Add to Cart
Request a Quote