Learn More
Immudex DEXTRAMER HLA-A*0201 Peptide Pool Negative Control, 5 mL, 500 tests, 32 nM, APC fluorochrome
SDP
Catalog No. NC3667580
Encompass
The Immudex DEXTRAMER HLA-A*0201 Peptide Pool Negative Control is a sophisticated reagent used for the identification and enumeration of antigen-specific T cells. Utilizing Dextramer technology, this reagent enhances binding affinity to T-cell receptors, ensuring a more reliable detection of low-affinity T cells while suppressing background staining. This product is essential in flow cytometry for distinguishing antigen-specific T cell staining.
  • Robust identification and enumeration of T cells
  • Enhances binding affinity to T-cell receptors
  • Reduces background staining for clearer results
  • Crucial for accurate flow cytometry thresholds
  • Applicable in immune-oncology, transplantation, autoimmunity, and infectious disease research
  • Manufactured under ISO 13485 and 21 CFR 820 quality standards
Catalog No. NC3667580 Supplier Immudex Supplier No. WB05535 AP 500
Add to Cart
Add to Cart
Request a Quote
The Immudex DEXTRAMER HLA-A*0201 Peptide Pool Negative Control is a sophisticated reagent used for the identification and enumeration of antigen-specific T cells. Utilizing Dextramer technology, this reagent enhances binding affinity to T-cell receptors, ensuring a more reliable detection of low-affinity T cells while suppressing background staining. This product is essential in flow cytometry for distinguishing antigen-specific T cell staining.
  • Robust identification and enumeration of T cells
  • Enhances binding affinity to T-cell receptors
  • Reduces background staining for clearer results
  • Crucial for accurate flow cytometry thresholds
  • Applicable in immune-oncology, transplantation, autoimmunity, and infectious disease research
  • Manufactured under ISO 13485 and 21 CFR 820 quality standards