Learn More
Medchemexpress LLC Dextran 20 | 9004-54-0 | 16000-24000 | 1 G
SDP
Supplier:  Medchemexpress LLC HY112624U1G
Encompass_Preferred
Dextran T20 is a dehydrated glucose polymer with an average molecular weight of 20,000. It has excellent biodegradability and good biocompatibility.
  • Excellent biodegradability and good biocompatibility
  • Can be used as a gene delivery vector
  • Can be used as an immune adjuvant carrier
  • Can be used as a hemoglobin stabilizer
  • Reduces hemoglobin self-oxidation rate by enhancing tetramer stability and oxygen affinity when bound to bovine hemoglobin
  • Enhances uptake by antigen-presenting cells and triggers a strong immune response when used to construct cytosine-phosphate-guanine (CpG)-right glycolic acid conjugates
  • Forms a dextran-peptide hybrid for safe and effective gene delivery (plasmid DNA) when bound to a cationic peptide R5H5
Catalog No. 50-004-37435