Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dextran, 250,000, Powder, Spectrum™ Chemical
SDP

Catalog No. 18602083 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-083 25 g Poly Bottle
18-602-084 100 g Poly Bottle
18-602-085 1 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-602-083 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. DE13025GM
Add to Cart
Add to Cart

DE130, 9004-54-0

Spectrum™ Chemical Dextran, 250,000, Powder is a complex branched glucan that is synthesized by certain lactic acid bacteria from sucrose. Commonly used in microsurgery to decrease the risk of vascular thrombosis, these agents are also used as effective osmotic agents and can treat hypovolemia. In the laboratory, Dextran is used to apply osmotic pressure to biological molecules and can, when applied with a fluorescent molecule, be used to create concentration gradients of diffusible molecules.

Chemical Identifiers

CAS 9004-54-0
Molecular Formula (C6H9O5)n(C6H10O5)m
Molecular Weight (g/mol) 504.44
MDL Number MFCD00130935
InChI Key FZWBNHMXJMCXLU-UHFFFAOYNA-N
IUPAC Name Dextran
SMILES O[C@@H]1[C@@H](O)[C@@H](-*)O[C@H](CO[C@H]2O[C@H](CO-*)[C@@H](O)[C@H](O-*)[C@H]2O)[C@H]1O

Specifications

Color White
CAS Max % 100%
Ash 2%
Quantity 25 g
Grade Reagent
Loss on Drying 10%
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.