Learn More
Selleck Chemical LLC Dextran S4492-100mg
SDP
Catalog No. 503110460
Encompass_Preferred
Dextran (Dextran 40 Macrodex) is a kind of biocompatible nontoxic and nonimmunogenic biological substance that has been widely used in drug-delivery systems
Catalog No. 50-311-0460 Supplier Selleck Chemical LLC Supplier No. S4492100MG
Dextran (Dextran 40 Macrodex) is a kind of biocompatible nontoxic and nonimmunogenic biological substance that has been widely used in drug-delivery systems