Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dextran 40 (Mw.=ca. 40,000), TCI America™
SDP

Catalog No. D1448500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1448500G 500 g
D144825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1448500G Supplier TCI America Supplier No. D1448500G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 9004-54-0
Molecular Formula (C6H9O5)n(C6H10O5)m
Molecular Weight (g/mol) 504.44
MDL Number MFCD00130935
InChI Key FZWBNHMXJMCXLU-UHFFFAOYNA-N
Synonym dextran, dextran, mw-86.000 aver., 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-2-yl oxy methyl tetrahydro-2h-pyran-2-yl oxy hexanal, 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxymethyl oxan-2-yl oxyhexanal, 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxymethyl tetrahydropyran-2-yl oxy-hexanal
PubChem CID 4125253
IUPAC Name Dextran
SMILES O[C@@H]1[C@@H](O)[C@@H](-*)O[C@H](CO[C@H]2O[C@H](CO-*)[C@@H](O)[C@H](O-*)[C@H]2O)[C@H]1O

Specifications

Color White
Physical Form Crystal-Powder at 20°C
Quantity 500 g
Chemical Name or Material Dextran 40 (Mw.=ca. 40,000)
RTECSNumber HH9246000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.