Learn More
LUNA NANOTECH INC Dextran Clio Mag Nanoparticles 40 Kda
Catalog No. NC3593169
Encompass_Preferred
Dextran Size: 40 kDa
Volume (@ 5 mg/mL Fe): 2 mL
General Information:
Streptavidin functionalized Dextran coated crosslinked iron oxide nanoparticles (CLIO-STRP) have 1-3 superparamagnetic Fe3O4 iron oxide cores (8-10 nm in diameter) imbedded within the matrix of dextran strands of 40 or 70 kDa in size. Dextran strands are crosslinked to prevent their disassociation, ensuring long-term biological stability. Streptavidin is covalently conjugated to the nanoparticle.
 Features:
Highly magnetic
Highly biocompatible, non-toxic
Long term stable under physiological conditions
40 kDa and 70 kDa dextran coats are available
Easy loading of biomolecules through Streptavidin-Biotin interaction
Applications:
In vitro and in vivo magnetic labels for cells and tissues
Magnetic cell separation
Probes for MRI
Specifications:
Iron Oxide Core Diameter: 8 &ndash
Catalog No. NC3593169 Supplier LUNA NANOTECH INC Supplier No. CLIOSTRP402
Dextran Size: 40 kDa
Volume (@ 5 mg/mL Fe): 2 mL
General Information:
Streptavidin functionalized Dextran coated crosslinked iron oxide nanoparticles (CLIO-STRP) have 1-3 superparamagnetic Fe3O4 iron oxide cores (8-10 nm in diameter) imbedded within the matrix of dextran strands of 40 or 70 kDa in size. Dextran strands are crosslinked to prevent their disassociation, ensuring long-term biological stability. Streptavidin is covalently conjugated to the nanoparticle.
 Features:
Highly magnetic
Highly biocompatible, non-toxic
Long term stable under physiological conditions
40 kDa and 70 kDa dextran coats are available
Easy loading of biomolecules through Streptavidin-Biotin interaction
Applications:
In vitro and in vivo magnetic labels for cells and tissues
Magnetic cell separation
Probes for MRI
Specifications:
Iron Oxide Core Diameter: 8 &ndash