Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dextran from Leuconostoc mesenteroides, For GPC, Mw 12,000, MilliporeSigma™ Supelco™

Catalog No. 111015813 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5813 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5813 Supplier MilliporeSigma Supelco Supplier No. 31418100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Analytical standard suitable for column calibration in gel permeation chromatography.

Food and Beverages
TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00130935

Specifications

Physical Form Neat
CAS 9004-54-0
Quantity 100 mg
Linear Formula (C6H10O5)n
Grade GPC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.