Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dextran, MP Biomedicals™

Catalog No. MP218014091 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP218014091 1 kg
MP218014010 10 g
MP218014080 100 g
MP218014090 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MP218014091 Supplier MP Biomedicals, Inc Supplier No. 0218014091
Add to Cart
Add to Cart

Improves performance of immobilized proteins acting on macromolecules. Dextran, MP Biomedicals is a branched polysaccharide made up of linear α (1→6) linked glucose units and α (1→3) link initiated branches. It ranges in size from 10

Depending upon its molecular weight it has different uses in different fields.

  • May be used as long hydrophilic spacer arms to improve the freedom of movement of conjugated/bound proteins.
  • May be derivatized for use in biosensor systems
  • Other applications include: platelet aggregant, plasma volume extender, osmotic pressure regulator, stabilizer, organ separation media, matrix component, copolymer, microcarrier, binding agent, viscosity modifier, antithrombotic, lubricant and physical structure component.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9004-54-0
Molecular Formula (C6H9O5)n(C6H10O5)m
Molecular Weight (g/mol) 504.44
MDL Number MFCD00130935
InChI Key FZWBNHMXJMCXLU-UHFFFAOYNA-N
Synonym dextran, dextran, mw-86.000 aver., 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-2-yl oxy methyl tetrahydro-2h-pyran-2-yl oxy hexanal, 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxymethyl oxan-2-yl oxyhexanal, 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxymethyl tetrahydropyran-2-yl oxy-hexanal
PubChem CID 4125253
IUPAC Name 2,3,4,5-tetrahydroxy-6-[3,4,5-trihydroxy-6-[[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxymethyl]oxan-2-yl]oxyhexanal
SMILES O[C@@H]1[C@@H](O)[C@@H](-*)O[C@H](CO[C@H]2O[C@H](CO-*)[C@@H](O)[C@H](O-*)[C@H]2O)[C@H]1O

Specifications

Physical Form Powder
Solubility Information Very soluble in water (50mg/mL), slightly soluble in alcohol.
Quantity 1 kg
Formula Weight 60,000-90,000
Grade Clinical
Recommended Storage Store at Room Temperature (15-30°C).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.