Learn More
Dextran, MW ca 500,000
Description
It is used in increasing the electronegativity and reducing erythrocyte aggregation and platelet adhesiveness and also serves as a plasminogen activator.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- Dextran is a glucose polymer biosynthesized by the non-pathogenic Leuconostoc mesenteroides
Application
- Dextran is a complex branched glucan that can be used to enhance electronegativity and reduce erythrocyte aggregation and platelet adhesiveness (anticoagulant activity)
- This compound promotes the activation of plasminogen by inhibiting α-2 antiplasmin
- Dextran can be used to reduce thrombosis in animal models
- It is used in osmotic stress techniques where osmotic pressure is applied to biological molecules
- Dextran can be used as an immobilization agent in biosensors
Specifications
Specifications
| Color | White |
| Physical Form | Powder |
| Merck Index | 14,2948 |
| Solubility Information | Soluble in water,dimethyl sulfoxide,ethylene glycol and glycerol. |
| Quantity | 100 g |
| Formula Weight | ≈500 kd |
| Chemical Name or Material | Dextran |
Safety and Handling
| EINECSNumber | 232-677-5 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.