Learn More
Bioworld DEXTRAN SEPAROPOREAGAROSE
Catalog No. 50197666
Encompass
DEXTRAN SEPAROPORE (AGAROSE) 4B-CL: Dextran coupled to a high strength Separopore 4B-CL matrix. Coupling Dextran to this material expands the surface area of the Separopore beads allowing for further binding of macromolecules.
Catalog No. 50-197-666 Supplier Bioworld Supplier No. 201810051
DEXTRAN SEPAROPORE (AGAROSE) 4B-CL: Dextran coupled to a high strength Separopore 4B-CL matrix. Coupling Dextran to this material expands the surface area of the Separopore beads allowing for further binding of macromolecules.