Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dextran Solution from Leuconostoc mesenteroides, Sigma-Aldrich

Catalog No. 01258960 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-960 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-960 Supplier MilliporeSigma Supelco Supplier No. D880250ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sterile dextran solution for use in hematology and histology applications.

Dextrans are used in many applications as platelet aggregants, plasma volume extenders, osmotic pressure regulators, stabilizers, organ separation media, matrix components, copolymers, microcarriers, binding agents, viscosity modifiers, antithrombotics, lubricants, and physical structure components. Dextran from Leuconostoc mesenteroides (Mw: 670,000) may be used as an analytical standard to calibrate the column for gel permeation chromatography (GPC). It has been used to sediment erythrocytes from blood along with 0. 9% sodium chloride.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9004-54-0
Molecular Weight (g/mol) 500000

Specifications

Quantity 50 mL
Physical Form Liquid
Chemical Name or Material Dextran
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.