Learn More
Research Products International Corp Dextran Sulfate, 1 KG
SDP
Catalog No. 501976892
Encompass_Preferred
Produced from Dextran 500,000 MW. Tested for use in nucleic acid hybridizations.
Free sulfate content: <0.02%
Catalog No. 50-197-6892 Supplier Research Products International Corp Supplier No. D200201000.0
Produced from Dextran 500,000 MW. Tested for use in nucleic acid hybridizations.
Free sulfate content: <0.02%