Learn More
VWR INTERNATIONAL LLC Dextran sulfate sodium salt, 9011-18-1, MFCD00081551, 250G
Supplier:  VWR INTERNATIONAL LLC 97061194
Encompass
Formula: (C₆H₇Na₃O₁₄S₃)n, MW: 500000 g/mol, Storage Temperature: Refrigerator, Ultra Pure Grade.
Catalog No. NC9748226