Learn More
Sigma Aldrich Fine Chemicals Biosciences Dextran Sulfate 50 Solutio
Catalog No. 502904557
Encompass
Dextran Sulfate (MW 500 000) is used as an additive in probe hybridization SOLUTIONons. Its presence accelerates the rate of hybridization by 10 fold. The addition of Dextran Sulfate favors the formation of probe networks resulting in greater sensitivity during membrane-bound nucleic acid analysis. The product is supplied as a 50% (w/v) liquid.
Catalog No. 50-290-4557 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. S4031
Dextran Sulfate (MW 500 000) is used as an additive in probe hybridization SOLUTIONons. Its presence accelerates the rate of hybridization by 10 fold. The addition of Dextran Sulfate favors the formation of probe networks resulting in greater sensitivity during membrane-bound nucleic acid analysis. The product is supplied as a 50% (w/v) liquid.